Product Details

SNP ID
rs16940244
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:109451883 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTCTGCCAACACCTGGACTTTAAG[C/G]AGGGCCGCCAGGCTAAATCAGGCCC
Phenotype
MIM: 613421 MIM: 614636
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KCTD10 PubMed Links

Gene Details

Gene
KCTD10
Gene Name
potassium channel tetramerization domain containing 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317395.1 Intron NP_001304324.1
NM_001317399.1 Intron NP_001304328.1
NM_031954.4 Intron NP_114160.1
Gene
MYO1H
Gene Name
myosin IH
There are no transcripts associated with this gene.

View Full Product Details