Product Details

SNP ID
rs17036290
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:37379670 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGTGCCATATCCCCTAACACTCT[A/T]AGTCGCAGTGCACGGGTCAATATTC
Phenotype
MIM: 611429
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
C3orf35 PubMed Links

Gene Details

Gene
C3orf35
Gene Name
chromosome 3 open reading frame 35
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302831.1 177 Intron NP_001289760.1
NM_001302832.1 177 Intron NP_001289761.1
NM_178339.2 177 Intron NP_848029.2
NM_178342.2 177 Intron NP_848032.1
NM_178344.2 177 Intron NP_848034.2
XM_017006291.1 177 UTR 5 XP_016861780.1
XM_017006292.1 177 UTR 5 XP_016861781.1
XM_017006293.1 177 Intron XP_016861782.1
XM_017006294.1 177 Intron XP_016861783.1
XM_017006295.1 177 Intron XP_016861784.1
XM_017006296.1 177 Intron XP_016861785.1

View Full Product Details