Product Details

SNP ID
rs17590869
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:68184985 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTAAGAATCTGGACTGTGGTGCTG[A/G]ACAGCCTAGGTTTGTATACCAGCTA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FTLP10 PubMed Links

Gene Details

Gene
FTLP10
Gene Name
ferritin light chain pseudogene 10
There are no transcripts associated with this gene.

Gene
TMPRSS11BNL
Gene Name
TMPRSS11B N-terminal like, pseudogene
There are no transcripts associated with this gene.

View Full Product Details