Product Details

SNP ID
rs17037911
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:107618987 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTTCCTTAGGAGGTCCACTTATTG[C/G]CTCCCTAAATAAGCTTCATTTTGAA
Phenotype
MIM: 603262
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
PAPSS1 PubMed Links

Gene Details

Gene
PAPSS1
Gene Name
3'-phosphoadenosine 5'-phosphosulfate synthase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005443.4 Intron NP_005434.4
XM_011532400.1 Intron XP_011530702.1
XM_011532401.1 Intron XP_011530703.1

View Full Product Details