Product Details
- SNP ID
-
rs17111008
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:150195230 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GCTTCGTGATAAGGAGCTGTATCTG[A/C]CTGCCGATGATCACTGCACTTTCAG
- Phenotype
-
MIM: 606205
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SLC6A7
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs138122237,rs75427098] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SLC6A7
- Gene Name
- solute carrier family 6 member 7
View Full Product Details