Product Details
- SNP ID
-
rs17110632
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:149858124 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGTAGATTAAAAGAGACTTCAGAAA[C/G]TTAACCATGGCTATGTGAGGTGATG
- Phenotype
-
MIM: 180071
MIM: 608886
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
PDE6A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78827133] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PDE6A
- Gene Name
- phosphodiesterase 6A
- Gene
- PPARGC1B
- Gene Name
- PPARG coactivator 1 beta
View Full Product Details