Product Details

SNP ID
rs16846830
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:196198388 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGAAGCCATTATTGAATCTCCCAGA[C/T]GGAAAAAGCTGTTCTCTTCTCTCTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HECW2 PubMed Links

Gene Details

Gene
HECW2
Gene Name
HECT, C2 and WW domain containing E3 ubiquitin protein ligase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304840.1 7828 Intron NP_001291769.1
NM_020760.2 7828 Intron NP_065811.1
XM_006712646.3 7828 UTR 3 XP_006712709.1
XM_006712648.3 7828 UTR 3 XP_006712711.1
XM_017004563.1 7828 UTR 3 XP_016860052.1

View Full Product Details