Product Details
- SNP ID
-
rs17318955
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:10559644 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ACTGATGGCTGGACGACATGCATGT[C/G]CTGAAATAGAAAAGATTGGGCTGAA
- Phenotype
-
MIM: 605702
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
LYVE1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs139395742] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- LYVE1
- Gene Name
- lymphatic vessel endothelial hyaluronan receptor 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006691.3 |
|
Intron |
|
|
NP_006682.2 |
- Gene
- MRVI1-AS1
- Gene Name
- MRVI1 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details