Product Details

SNP ID
rs11556024
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:116748335 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TACCTCAGTCCAAACATCAGGTCTC[A/G]AATATTCTTCTGTGGTCAAAACTGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BUD13 PubMed Links

Gene Details

Gene
BUD13
Gene Name
BUD13 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159736.1 2615 UTR 3 NP_001153208.1
NM_032725.3 2615 UTR 3 NP_116114.1
XM_011543035.2 2615 UTR 3 XP_011541337.1

View Full Product Details