Product Details

SNP ID
rs17832261
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:110902356 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTACAGTCAAGATTCTGCTATGCT[C/G]TGTGTTCAAGGTGTTCTGGACTAAG
Phenotype
MIM: 613605 MIM: 608610
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
BBIP1 PubMed Links

Gene Details

Gene
BBIP1
Gene Name
BBSome interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195304.1 Intron NP_001182233.1
NM_001195305.1 Intron NP_001182234.1
NM_001195306.1 Intron NP_001182235.1
NM_001195307.1 Intron NP_001182236.1
NM_001243783.1 Intron NP_001230712.1
Gene
MIR4680
Gene Name
microRNA 4680
There are no transcripts associated with this gene.

Gene
PDCD4
Gene Name
programmed cell death 4 (neoplastic transformation inhibitor)
There are no transcripts associated with this gene.

View Full Product Details