Product Details

SNP ID
rs17117493
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:104644838 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGATATGAGTTCCCTTTTGGGGAG[C/T]GTTTTCAGTAACCCTATGAAATTGA
Phenotype
MIM: 606285
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SORCS3 PubMed Links

Gene Details

Gene
SORCS3
Gene Name
sortilin related VPS10 domain containing receptor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014978.2 Intron NP_055793.1
XM_011539542.2 Intron XP_011537844.1

View Full Product Details