Product Details

SNP ID
rs16846879
Assay Type
Functionally tested
NCBI dbSNP Submissions
32
Location
Chr.1:200553367 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCTTTTTCTTTCATGGGTGGTCATA[A/G]AAGTGCCTACACATCAGTATTCACA
Phenotype
MIM: 611279
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIF14 PubMed Links

Gene Details

Gene
KIF14
Gene Name
kinesin family member 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001305792.1 4719 UTR 3 NP_001292721.1
NM_014875.2 4719 UTR 3 NP_055690.1
XM_011510231.2 4719 UTR 3 XP_011508533.1
XM_011510232.2 4719 UTR 3 XP_011508534.1
XM_011510233.2 4719 UTR 3 XP_011508535.1
XM_011510235.2 4719 UTR 3 XP_011508537.1
XM_011510236.1 4719 UTR 3 XP_011508538.1
XM_017003005.1 4719 UTR 3 XP_016858494.1
XM_017003006.1 4719 UTR 3 XP_016858495.1
XM_017003007.1 4719 UTR 3 XP_016858496.1

View Full Product Details