Product Details

SNP ID
rs17854938
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:130173675 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGACGCTGGTAGATGAGCTCATAA[C/T]GGGGAGGGTTGATAGCACTCTGATA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMEM209 PubMed Links

Gene Details

Gene
TMEM209
Gene Name
transmembrane protein 209
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301163.1 1927 Missense Mutation CAT,CGT H463R NP_001288092.1
NM_032842.3 1927 Missense Mutation CAT,CGT H505R NP_116231.2
XM_005250655.1 1927 Missense Mutation CAT,CGT H504R XP_005250712.1
XM_011516646.2 1927 Missense Mutation CAT,CGT H504R XP_011514948.1

View Full Product Details