Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001135914.1 | Intron | NP_001129386.1 | |||
NM_199349.2 | Intron | NP_955381.2 | |||
XM_017012184.1 | Intron | XP_016867673.1 | |||
XM_017012185.1 | Intron | XP_016867674.1 | |||
XM_017012186.1 | Intron | XP_016867675.1 | |||
XM_017012187.1 | Intron | XP_016867676.1 | |||
XM_017012188.1 | Intron | XP_016867677.1 | |||
XM_017012189.1 | Intron | XP_016867678.1 | |||
XM_017012190.1 | Intron | XP_016867679.1 | |||
XM_017012191.1 | Intron | XP_016867680.1 | |||
XM_017012192.1 | Intron | XP_016867681.1 | |||
XM_017012193.1 | Intron | XP_016867682.1 | |||
XM_017012194.1 | Intron | XP_016867683.1 |