Product Details

SNP ID
rs17775910
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:134567887 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACTTTCCTTCCTTTCTTTTGAAAA[C/T]TTTTCTAAACTGTGTATCCTCCCAG
Phenotype
MIM: 616336
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AKR1B15 PubMed Links

Gene Details

Gene
AKR1B15
Gene Name
aldo-keto reductase family 1 member B15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080538.2 Intron NP_001074007.2
XM_011516241.2 Intron XP_011514543.2
XM_017012224.1 Intron XP_016867713.1

View Full Product Details