Product Details

SNP ID
rs16979925
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:15500076 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCTGCCAACCCAAGACTTCTTCAC[A/G]TCATAGATACTTACAAATAAGTTAC
Phenotype
MIM: 300335 MIM: 300101 MIM: 300931
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACE2 PubMed Links

Gene Details

Gene
ACE2
Gene Name
angiotensin I converting enzyme 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021804.2 Intron NP_068576.1
XM_011545549.1 Intron XP_011543851.1
XM_011545551.2 Intron XP_011543853.1
XM_011545552.1 Intron XP_011543854.1
Gene
BMX
Gene Name
BMX non-receptor tyrosine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320866.1 Intron NP_001307795.1
NM_001721.6 Intron NP_001712.1
NM_203281.2 Intron NP_975010.1
XM_017029752.1 Intron XP_016885241.1
Gene
PIR
Gene Name
pirin
There are no transcripts associated with this gene.

Gene
PIR-FIGF
Gene Name
PIR-FIGF readthrough
There are no transcripts associated with this gene.

View Full Product Details