Product Details

SNP ID
rs17246987
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:18246375 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTCTATCAGAGACATGGTTGGTGAT[A/C]CAGATCCCTAGAGACAGTTCTGTGC
Phenotype
MIM: 300208
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SCML2 PubMed Links

Gene Details

Gene
SCML2
Gene Name
sex comb on midleg-like 2 (Drosophila)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006089.2 Intron NP_006080.1
XM_006724458.1 Intron XP_006724521.1
XM_017029219.1 Intron XP_016884708.1
XM_017029220.1 Intron XP_016884709.1
XM_017029221.1 Intron XP_016884710.1
XM_017029222.1 Intron XP_016884711.1

View Full Product Details