Product Details

SNP ID
rs7117020
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:57732183 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTTTTCTCATGTCAATCTGCCTTT[G/T]GTCAGCTCATTTTCAGCAAAACTTT
Phenotype
MIM: 607914 MIM: 616715
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C11orf31 PubMed Links

Gene Details

Gene
C11orf31
Gene Name
chromosome 11 open reading frame 31
There are no transcripts associated with this gene.

Gene
TMX2
Gene Name
thioredoxin related transmembrane protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144012.2 Intron NP_001137484.1
NM_015959.3 Intron NP_057043.1
Gene
TMX2-CTNND1
Gene Name
TMX2-CTNND1 readthrough (NMD candidate)
There are no transcripts associated with this gene.

View Full Product Details