Product Details

SNP ID
rs17144453
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:82981089 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TACTAAAAAATCTACAAAATCCTAA[A/G]CCTTGGTCTGTCCAAGATGGAGAGT
Phenotype
MIM: 605693
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RAB30 PubMed Links

Gene Details

Gene
RAB30
Gene Name
RAB30, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286059.1 1827 UTR 3 NP_001272988.1
NM_001286060.1 1827 UTR 3 NP_001272989.1
NM_001286061.1 1827 UTR 3 NP_001272990.1
NM_014488.4 1827 UTR 3 NP_055303.2

View Full Product Details