Product Details

SNP ID
rs17103147
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:35278712 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTGGAGGTGGCTATGGCAGGTCTT[C/T]GGAGAGGTAAGTCCCTCACTCAGAC
Phenotype
MIM: 609947 MIM: 602855
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIAA0391 PubMed Links

Gene Details

Gene
KIAA0391
Gene Name
KIAA0391
There are no transcripts associated with this gene.

Gene
PSMA6
Gene Name
proteasome subunit alpha 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282232.1 Intron NP_001269161.1
NM_001282233.1 Intron NP_001269162.1
NM_001282234.1 Intron NP_001269163.1
NM_002791.2 Intron NP_002782.1
XM_017021468.1 Intron XP_016876957.1

View Full Product Details