Product Details

SNP ID
rs17097482
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:30900867 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATAAATAGGTGAAATTTGGAAACAA[A/T]TTGAAAGACTATATGCTGCTTATTT
Phenotype
MIM: 614766
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
STRN3 PubMed Links

Gene Details

Gene
STRN3
Gene Name
striatin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001083893.1 Intron NP_001077362.1
NM_014574.3 Intron NP_055389.3
XM_005267569.3 Intron XP_005267626.1
XM_005267570.3 Intron XP_005267627.1

View Full Product Details