Product Details
- SNP ID
-
rs17850149
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:21211482 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCAGCAGAGTCATCTGCACCCCCC[C/T]CAGACTCCATCTTCACATTAGTCTC
- Phenotype
-
MIM: 164020
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
HNRNPC
PubMed Links
Gene Details
- Gene
- HNRNPC
- Gene Name
- heterogeneous nuclear ribonucleoprotein C (C1/C2)
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001077442.1 |
966 |
Missense Mutation |
GAG,GGG |
E254G |
NP_001070910.1 |
NM_001077443.1 |
966 |
Missense Mutation |
GAG,GGG |
E241G |
NP_001070911.1 |
NM_004500.3 |
966 |
Missense Mutation |
GAG,GGG |
E241G |
NP_004491.2 |
NM_031314.2 |
966 |
Missense Mutation |
GAG,GGG |
E254G |
NP_112604.2 |
XM_006720125.2 |
966 |
Missense Mutation |
GAG,GGG |
E241G |
XP_006720188.1 |
XM_011536708.1 |
966 |
Missense Mutation |
GAG,GGG |
E254G |
XP_011535010.1 |
XM_011536709.2 |
966 |
Missense Mutation |
GAG,GGG |
E254G |
XP_011535011.1 |
XM_011536710.2 |
966 |
Missense Mutation |
GAG,GGG |
E254G |
XP_011535012.1 |
XM_011536711.2 |
966 |
Missense Mutation |
GAG,GGG |
E254G |
XP_011535013.1 |
XM_011536712.1 |
966 |
Missense Mutation |
GAG,GGG |
E241G |
XP_011535014.1 |
XM_017021252.1 |
966 |
Missense Mutation |
GAG,GGG |
E241G |
XP_016876741.1 |
XM_017021253.1 |
966 |
Missense Mutation |
GAG,GGG |
E241G |
XP_016876742.1 |
- Gene
- LINC00641
- Gene Name
- long intergenic non-protein coding RNA 641
There are no transcripts associated with this gene.
View Full Product Details