Product Details

SNP ID
rs17004729
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:45071579 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGGAGACAGTCACAGGGTCATGC[C/T]ATTTAACTGCACTGAAGGGAAAACG
Phenotype
MIM: 601218
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADARB1 PubMed Links

Gene Details

Gene
ADARB1
Gene Name
adenosine deaminase, RNA specific B1
There are no transcripts associated with this gene.

Gene
LOC105372836
Gene Name
uncharacterized LOC105372836
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011529812.2 Intron XP_011528114.1
Gene
SSR4P1
Gene Name
signal sequence receptor subunit 4 pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details