Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_007031.1 | Intron | NP_008962.1 | |||
XM_011529443.2 | Intron | XP_011527745.2 | |||
XM_011529445.2 | Intron | XP_011527747.2 | |||
XM_011529446.2 | Intron | XP_011527748.1 | |||
XM_011529447.2 | Intron | XP_011527749.1 | |||
XM_017028267.1 | Intron | XP_016883756.1 | |||
XM_017028268.1 | Intron | XP_016883757.1 | |||
XM_017028269.1 | Intron | XP_016883758.1 | |||
XM_017028270.1 | Intron | XP_016883759.1 | |||
XM_017028271.1 | Intron | XP_016883760.1 | |||
XM_017028272.1 | Intron | XP_016883761.1 |