Product Details
- SNP ID
-
rs16898614
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:31056063 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGTGGAATATGTTTTACCAGAATTA[A/G]ACCAAAAATGCCAAATGATCCCTAA
- Phenotype
-
MIM: 613918
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
HCG22
PubMed Links
Gene Details
- Gene
- HCG22
- Gene Name
- HLA complex group 22
There are no transcripts associated with this gene.
View Full Product Details