Product Details

SNP ID
rs72552760
Assay Type
DME
NCBI dbSNP Submissions
NA
Location
Chr.6:52899946 on Build GRCh38
Set Membership
DME Validated Inventoried
Context Sequence [VIC/FAM]
AGCTTCACCTACTTTTTCGAAGGCA[-/G]GGGAAATAGCGACTTTTTGTTTTCT
Phenotype
MIM: 605449
Polymorphism
-/G, Insertion/deletion
Allele Nomenclature
Literature Links
GSTA3 PubMed Links
Additional Information
For this assay, SNP(s) [rs45602042] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
GSTA3
Gene Name
glutathione S-transferase alpha 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000847.4 306 Frame Shift InDel CCC,CCT P134P NP_000838.3
XM_006715072.3 306 Frame Shift InDel CCC,CCT P84P XP_006715135.1
XM_011514533.2 306 Frame Shift InDel CCC,CCT P41P XP_011512835.1
XM_017010794.1 306 Frame Shift InDel CCC,CCT P84P XP_016866283.1

View Full Product Details