Product Details

SNP ID
rs17851798
Assay Type
DME
NCBI dbSNP Submissions
NA
Location
Chr.6:52902429 on Build GRCh38
Set Membership
HapMap DME Validated Inventoried
Context Sequence [VIC/FAM]
GAATGGCTCTGGTCTGTACCAACTT[A/C]ATCCCATCAATCTCAACCATTGGTA
Phenotype
MIM: 605449
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GSTA3 PubMed Links

Gene Details

Gene
GSTA3
Gene Name
glutathione S-transferase alpha 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000847.4 102 Missense Mutation NP_000838.3
XM_006715072.3 102 Missense Mutation XP_006715135.1
XM_011514533.2 102 Intron XP_011512835.1
XM_017010794.1 102 Missense Mutation XP_016866283.1

View Full Product Details