Product Details

SNP ID
rs16914034
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:59191027 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTACAGGAGCTACCATTCCTGTGAT[G/T]TTTTATGTTATCTATTTTCCTCAAT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PHYHIPL PubMed Links

Gene Details

Gene
PHYHIPL
Gene Name
phytanoyl-CoA 2-hydroxylase interacting protein like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001143774.1 Intron NP_001137246.1
NM_032439.3 Intron NP_115815.2
XM_011540275.2 Intron XP_011538577.1
XM_011540276.2 Intron XP_011538578.1
XM_017016782.1 Intron XP_016872271.1
XM_017016783.1 Intron XP_016872272.1

View Full Product Details