Product Details

SNP ID
rs17318866
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:3836964 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATTATCAAGAGAAGATACAGGCTTC[A/G]TGCAGGCTTTTGGAAAATCTGGGTG
Phenotype
MIM: 614686
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM50B PubMed Links

Gene Details

Gene
FAM50B
Gene Name
family with sequence similarity 50 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012135.2 Intron NP_036267.1
XM_017010729.1 Intron XP_016866218.1

View Full Product Details