Product Details

SNP ID
rs17339677
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:3072433 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGCGCTGAAACTTGATGGCTCC[A/G]AACACCCTCGAAGCGCGCCACTCGC
Phenotype
MIM: 167050
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OXT PubMed Links

Gene Details

Gene
OXT
Gene Name
oxytocin/neurophysin I prepropeptide
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000915.3 566 UTR 3 NP_000906.1
XM_011529238.1 566 UTR 3 XP_011527540.1

View Full Product Details