Product Details

SNP ID
rs17662001
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:26457082 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAATATAGTCTGCTATGTTGCAAAA[C/T]AGCTTACTAGATGAGATGGGAAAGC
Phenotype
MIM: 613420
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KCTD1 PubMed Links

Gene Details

Gene
KCTD1
Gene Name
potassium channel tetramerization domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136205.2 Intron NP_001129677.1
NM_001142730.2 Intron NP_001136202.1
NM_001258221.1 Intron NP_001245150.1
NM_001258222.1 Intron NP_001245151.1
NM_198991.3 Intron NP_945342.1
XM_017025708.1 Intron XP_016881197.1

View Full Product Details