Product Details

SNP ID
hCV33461171
Assay Type
DME
NCBI dbSNP Submissions
NA
Location
Chr.7:95302279 on Build GRCh38
Set Membership
DME Validated Inventoried
Context Sequence [VIC/FAM]
TTGGGATGGCATCCAACCCAAAGGT[-/A]CTCCTGTCTCAGGATCCACAGATAT
Phenotype
MIM: 168820 MIM: 602468
Polymorphism
-/A, Insertion/deletion
Allele Nomenclature
Literature Links
PON1 PubMed Links

Gene Details

Gene
PON1
Gene Name
paraoxonase 1
There are no transcripts associated with this gene.

Gene
PPP1R9A
Gene Name
protein phosphatase 1 regulatory subunit 9A
There are no transcripts associated with this gene.

View Full Product Details