Product Details

SNP ID
rs16848367
Assay Type
Functionally tested
NCBI dbSNP Submissions
26
Location
Chr.1:228143438 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCGTATTAGAAAAGTCACCATAATA[A/C]CATCAAGTCATAATTTCTGTGATCT
Phenotype
MIM: 608803 MIM: 139270
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GJC2 PubMed Links

Gene Details

Gene
GJC2
Gene Name
gap junction protein gamma 2
There are no transcripts associated with this gene.

Gene
GUK1
Gene Name
guanylate kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000858.5 1586 Intron NP_000849.1
NM_001159390.1 1586 Intron NP_001152862.1
NM_001159391.1 1586 Intron NP_001152863.1
NM_001242839.1 1586 Intron NP_001229768.1
NM_001242840.1 1586 Intron NP_001229769.1
XM_005273103.3 1586 Intron XP_005273160.1
XM_005273104.4 1586 Intron XP_005273161.1
XM_005273105.3 1586 Intron XP_005273162.1
XM_005273106.2 1586 Intron XP_005273163.1
XM_011544174.2 1586 UTR 5 XP_011542476.1
XM_017001091.1 1586 Intron XP_016856580.1
XM_017001092.1 1586 Intron XP_016856581.1
XM_017001093.1 1586 Intron XP_016856582.1
XM_017001094.1 1586 Intron XP_016856583.1
XM_017001095.1 1586 Intron XP_016856584.1
XM_017001096.1 1586 Intron XP_016856585.1

View Full Product Details