Product Details

SNP ID
rs16959046
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:66308516 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGAGAAGTGACTATGTGCTTTCTAT[A/G]TAGTTCTTTGCTTTGGTGAAGAATT
Phenotype
MIM: 176960
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PRKCA PubMed Links

Gene Details

Gene
PRKCA
Gene Name
protein kinase C alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002737.2 Intron NP_002728.1
XM_017024836.1 Intron XP_016880325.1
XM_017024837.1 Intron XP_016880326.1
XM_017024838.1 Intron XP_016880327.1
XM_017024839.1 Intron XP_016880328.1
XM_017024840.1 Intron XP_016880329.1
XM_017024841.1 Intron XP_016880330.1

View Full Product Details