Product Details

SNP ID
rs17047577
Assay Type
Functionally tested
NCBI dbSNP Submissions
23
Location
Chr.1:218288025 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACAAAGTAATCTTCTGTTATGGCTC[C/T]AGCTGTTCAGAGAAGTGACTAGAAG
Phenotype
MIM: 611193
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RRP15 PubMed Links

Gene Details

Gene
RRP15
Gene Name
ribosomal RNA processing 15 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016052.3 Intron NP_057136.2
XM_011509597.2 Intron XP_011507899.1

View Full Product Details