Product Details

SNP ID
rs16875575
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:5435478 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TACTTGTGAAGGGCTATTGTGGTTA[C/T]GTTTAAAAGTAAGTCCTTATTTCTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ICE1 PubMed Links

Gene Details

Gene
ICE1
Gene Name
interactor of little elongation complex ELL subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015325.2 Intron NP_056140.1
XM_011513999.2 Intron XP_011512301.1
XM_017009285.1 Intron XP_016864774.1

View Full Product Details