Product Details

SNP ID
rs17638694
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12165463 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGAAGATAATGTATTTCCCAAGAG[A/G]TAGATTTCAGGGGGAAAGAGATTAG
Phenotype
MIM: 604078
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF136 PubMed Links

Gene Details

Gene
ZNF136
Gene Name
zinc finger protein 136
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003437.3 Intron NP_003428.1
XM_011528269.2 Intron XP_011526571.1
XM_017027243.1 Intron XP_016882732.1
Gene
ZNF625
Gene Name
zinc finger protein 625
There are no transcripts associated with this gene.

Gene
ZNF625-ZNF20
Gene Name
ZNF625-ZNF20 readthrough (NMD candidate)
There are no transcripts associated with this gene.

View Full Product Details