Product Details

SNP ID
rs17545743
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:62371931 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGTTTCTTACTATAATGCTTCAGTG[A/T]TGTTGTCAGAGAGTTTCCATTCAGC
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
EFCAB3 PubMed Links

Gene Details

Gene
EFCAB3
Gene Name
EF-hand calcium binding domain 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144933.1 Intron NP_001138405.1
NM_173503.3 Intron NP_775774.1
XM_011524380.1 Intron XP_011522682.1
XM_011524381.1 Intron XP_011522683.1

View Full Product Details