Product Details

SNP ID
rs17003955
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:109624924 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCCCATGCCCATGCTGGGACGAG[A/G]GCCAGCGCCCAAGCCGCTGGCATTA
Phenotype
MIM: 300328
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCNE5 PubMed Links

Gene Details

Gene
KCNE5
Gene Name
potassium voltage-gated channel subfamily E regulatory subunit 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012282.2 241 Missense Mutation CCT,TCT P33S NP_036414.1

View Full Product Details