Product Details

SNP ID
rs16985810
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:109624243 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACGGCCCAGAGTTCTGATAGAAT[G/T]CATTGCTTCAGTTCAGTGCCAAGTG
Phenotype
MIM: 300328
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KCNE5 PubMed Links

Gene Details

Gene
KCNE5
Gene Name
potassium voltage-gated channel subfamily E regulatory subunit 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012282.2 922 UTR 3 NP_036414.1

View Full Product Details