Product Details

SNP ID
rs17035853
Assay Type
Functionally tested
NCBI dbSNP Submissions
21
Location
Chr.1:116490008 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAATGTCCTCTTTCCATGCTAGTG[C/T]CCAGGTTTCCTCTCAGACAAGTAGC
Phenotype
MIM: 153420
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CD58 PubMed Links

Gene Details

Gene
CD58
Gene Name
CD58 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144822.1 15996 Intron NP_001138294.1
NM_001779.2 15996 Intron NP_001770.1
XM_017002869.1 15996 UTR 3 XP_016858358.1
Gene
LOC105378922
Gene Name
uncharacterized LOC105378922
There are no transcripts associated with this gene.

View Full Product Details