Product Details

SNP ID
rs35803472
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:43533803 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTATCTCTACTACTACAGCATCTC[C/T]TAAGCCAAACAGCCCCACCACAGAG
Phenotype
MIM: 600178 MIM: 610979
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAP1A PubMed Links

Gene Details

Gene
MAP1A
Gene Name
microtubule associated protein 1A
There are no transcripts associated with this gene.

Gene
PPIP5K1
Gene Name
diphosphoinositol pentakisphosphate kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001130858.2 5365 UTR 3 NP_001124330.1
NM_001130859.2 5365 UTR 3 NP_001124331.1
NM_001190214.1 5365 UTR 3 NP_001177143.1
NM_014659.5 5365 UTR 3 NP_055474.3
XM_005254803.1 5365 UTR 3 XP_005254860.1
XM_005254804.1 5365 UTR 3 XP_005254861.1
XM_006720786.2 5365 UTR 3 XP_006720849.1
XM_011522249.1 5365 UTR 3 XP_011520551.1
XM_011522250.1 5365 UTR 3 XP_011520552.1
XM_011522252.1 5365 UTR 3 XP_011520554.1
XM_011522254.1 5365 UTR 3 XP_011520556.1
XM_011522258.1 5365 UTR 3 XP_011520560.1
XM_011522259.1 5365 UTR 3 XP_011520561.1
XM_017022735.1 5365 UTR 3 XP_016878224.1
XM_017022736.1 5365 UTR 3 XP_016878225.1
XM_017022737.1 5365 UTR 3 XP_016878226.1
XM_017022738.1 5365 UTR 3 XP_016878227.1
XM_017022739.1 5365 UTR 3 XP_016878228.1
XM_017022740.1 5365 UTR 3 XP_016878229.1
XM_017022741.1 5365 UTR 3 XP_016878230.1
XM_017022742.1 5365 UTR 3 XP_016878231.1
XM_017022743.1 5365 UTR 3 XP_016878232.1
XM_017022744.1 5365 UTR 3 XP_016878233.1
XM_017022745.1 5365 UTR 3 XP_016878234.1
XM_017022746.1 5365 UTR 3 XP_016878235.1
XM_017022747.1 5365 UTR 3 XP_016878236.1
XM_017022748.1 5365 UTR 3 XP_016878237.1
XM_017022749.1 5365 UTR 3 XP_016878238.1
XM_017022750.1 5365 UTR 3 XP_016878239.1
XM_017022751.1 5365 UTR 3 XP_016878240.1
XM_017022752.1 5365 UTR 3 XP_016878241.1
XM_017022753.1 5365 UTR 3 XP_016878242.1
XM_017022754.1 5365 UTR 3 XP_016878243.1
XM_017022755.1 5365 UTR 3 XP_016878244.1
XM_017022756.1 5365 UTR 3 XP_016878245.1
XM_017022757.1 5365 UTR 3 XP_016878246.1
XM_017022758.1 5365 UTR 3 XP_016878247.1
XM_017022759.1 5365 Intron XP_016878248.1
XM_017022760.1 5365 UTR 3 XP_016878249.1

View Full Product Details