Product Details
- SNP ID
-
rs16978840
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:44213771 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGGGAATGTAGGTATGGATTTTACT[G/T]TGGAAGACCTCAAAAACTGCACTGA
- Phenotype
-
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ZNF227
PubMed Links
Gene Details
- Gene
- ZNF227
- Gene Name
- zinc finger protein 227
View Full Product Details