Product Details

SNP ID
rs35304389
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:52115236 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAAGTCTAAGATGGACACGCTGA[C/G]TAAAGGAATTCCCACACTGATTGCA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF616 PubMed Links

Gene Details

Gene
ZNF616
Gene Name
zinc finger protein 616
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178523.4 2211 Missense Mutation ACT,AGT T643S NP_848618.2
XM_017027438.1 2211 Missense Mutation ACT,AGT T674S XP_016882927.1

View Full Product Details