Product Details

SNP ID
rs17694981
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:51764333 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCGGCTGAAAATTTCACATCTAGTT[A/T]TTACCATTTTCTCATTATAATGTCA
Phenotype
MIM: 136538
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
FPR2 PubMed Links

Gene Details

Gene
FPR2
Gene Name
formyl peptide receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005738.1 Intron NP_001005738.1
NM_001462.3 Intron NP_001453.1
XM_006723120.3 Intron XP_006723183.1
XM_017026542.1 Intron XP_016882031.1

View Full Product Details