Product Details

SNP ID
rs16966134
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:40699363 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTATTACTCTCTATATATACAAGT[C/G]TAAGCCCTGAGTCCCATTTTGGTTT
Phenotype
MIM: 607742
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KRT24 PubMed Links

Gene Details

Gene
KRT24
Gene Name
keratin 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019016.2 Intron NP_061889.2
XM_006721739.3 Intron XP_006721802.1
XM_011524460.2 Intron XP_011522762.1
XM_017024299.1 Intron XP_016879788.1
Gene
LOC105371775
Gene Name
uncharacterized LOC105371775
There are no transcripts associated with this gene.

View Full Product Details