Product Details

SNP ID
rs17624067
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:37519867 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAAATGGCTACATCCTGAAGCTCT[C/T]TGATCTCCATCTGTGTGTCACTACA
Phenotype
MIM: 606618 MIM: 607291
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DUSP14 PubMed Links

Gene Details

Gene
DUSP14
Gene Name
dual specificity phosphatase 14
There are no transcripts associated with this gene.

Gene
SYNRG
Gene Name
synergin, gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163544.2 Intron NP_001157016.1
NM_001163545.2 Intron NP_001157017.1
NM_001163546.2 Intron NP_001157018.1
NM_001163547.2 Intron NP_001157019.1
NM_007247.5 Intron NP_009178.3
NM_080550.4 Intron NP_542117.3
NM_198882.2 Intron NP_942583.1
XM_005256980.4 Intron XP_005257037.1
XM_017024076.1 Intron XP_016879565.1
XM_017024077.1 Intron XP_016879566.1
XM_017024078.1 Intron XP_016879567.1
XM_017024079.1 Intron XP_016879568.1
XM_017024080.1 Intron XP_016879569.1
XM_017024081.1 Intron XP_016879570.1
XM_017024082.1 Intron XP_016879571.1
XM_017024083.1 Intron XP_016879572.1
XM_017024084.1 Intron XP_016879573.1
XM_017024085.1 Intron XP_016879574.1
XM_017024086.1 Intron XP_016879575.1
XM_017024087.1 Intron XP_016879576.1
XM_017024088.1 Intron XP_016879577.1
XM_017024089.1 Intron XP_016879578.1
XM_017024090.1 Intron XP_016879579.1
XM_017024091.1 Intron XP_016879580.1
XM_017024092.1 Intron XP_016879581.1
XM_017024093.1 Intron XP_016879582.1
XM_017024094.1 Intron XP_016879583.1
XM_017024095.1 Intron XP_016879584.1
XM_017024096.1 Intron XP_016879585.1
XM_017024097.1 Intron XP_016879586.1
XM_017024098.1 Intron XP_016879587.1
XM_017024099.1 Intron XP_016879588.1
XM_017024100.1 Intron XP_016879589.1
XM_017024101.1 Intron XP_016879590.1
XM_017024102.1 Intron XP_016879591.1
XM_017024103.1 Intron XP_016879592.1
XM_017024104.1 Intron XP_016879593.1
XM_017024105.1 Intron XP_016879594.1
XM_017024106.1 Intron XP_016879595.1

View Full Product Details