Product Details

SNP ID
rs16941273
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:46923841 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTGTCCTTTATTCGCATCGTCCTGC[A/G]CTTTCGGAATTGCTGAGATACGTAG
Phenotype
MIM: 604027
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GOSR2 PubMed Links

Gene Details

Gene
GOSR2
Gene Name
golgi SNAP receptor complex member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012511.2 Intron NP_001012529.1
NM_001321133.1 Intron NP_001308062.1
NM_001321134.1 Intron NP_001308063.1
NM_004287.4 Intron NP_004278.2
NM_054022.3 Intron NP_473363.1
XM_005257844.3 Intron XP_005257901.1
XM_006722190.3 Intron XP_006722253.1
XM_011525501.2 Intron XP_011523803.1
XM_011525502.2 Intron XP_011523804.1
XM_017025378.1 Intron XP_016880867.1
XM_017025379.1 Intron XP_016880868.1
XM_017025380.1 Intron XP_016880869.1
XM_017025381.1 Intron XP_016880870.1
XM_017025382.1 Intron XP_016880871.1
XM_017025383.1 Intron XP_016880872.1
XM_017025384.1 Intron XP_016880873.1
XM_017025385.1 Intron XP_016880874.1
XM_017025386.1 Intron XP_016880875.1
XM_017025387.1 Intron XP_016880876.1
XM_017025388.1 Intron XP_016880877.1
XM_017025389.1 Intron XP_016880878.1
XM_017025390.1 Intron XP_016880879.1
XM_017025391.1 Intron XP_016880880.1
XM_017025392.1 Intron XP_016880881.1
XM_017025393.1 Intron XP_016880882.1
XM_017025394.1 Intron XP_016880883.1
XM_017025395.1 Intron XP_016880884.1

View Full Product Details