Product Details

SNP ID
rs17354678
Assay Type
Functionally tested
NCBI dbSNP Submissions
31
Location
Chr.1:145745816 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAACATCTGTCTTTCTGTGCCTGTA[T/C]GCTGCCCAACTCATTCCCAAAAAGG
Phenotype
MIM: 604463
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
CD160 PubMed Links

Gene Details

Gene
CD160
Gene Name
CD160 molecule
There are no transcripts associated with this gene.

Gene
RNF115
Gene Name
ring finger protein 115
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014455.3 2263 Intron NP_055270.1
XM_005272952.4 2263 UTR 3 XP_005273009.1

View Full Product Details