Product Details

SNP ID
hCV33590543
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.1:145923556 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAATCTCCATTACTGGAAAGCAGG[T/C]ACTTTAACCTTGTAGCTCTAAACAG
Phenotype
MIM: 612875 MIM: 603867 MIM: 605313
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
GNRHR2 PubMed Links

Gene Details

Gene
GNRHR2
Gene Name
gonadotropin releasing hormone receptor 2 (pseudogene)
There are no transcripts associated with this gene.

Gene
LIX1L
Gene Name
limb and CNS expressed 1 like
There are no transcripts associated with this gene.

Gene
PEX11B
Gene Name
peroxisomal biogenesis factor 11 beta
There are no transcripts associated with this gene.

Gene
RBM8A
Gene Name
RNA binding motif protein 8A
There are no transcripts associated with this gene.

View Full Product Details